2-Amino-5-[4-(tert-butyl)phenyl]-1,3-thiazole-4-carbohydrazide ≥95%

Supplier: Matrix Scientific

043757-500MG 043757-1G 043757-5G
101862-416EA 278.46 USD
101862-416 101862-418 101862-420
2-Amino-5-[4-(tert-butyl)phenyl]-1,3-thiazole-4-carbohydrazide ≥95%
2-Amino-5-[4-(tert-butyl)phenyl]-1,3-thiazole-4-carbohydrazide

Hazard: Irritant

Formula: C₁₄H₁₈N₄OS
Melting Pt: 170 °C
MDL Number: MFCD06200938
CAS Number: 886361-56-4

Order Now

Specification Test Results

Melting point 170°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR