2-Amino-5-(4-chlorophenyl)-4-thiazolecarbohydrazide ≥95%

Supplier: Matrix Scientific

043756-500MG 043756-1G 043756-5G
101862-410EA 278.46 USD
101862-410 101862-412 101862-414
2-Amino-5-(4-chlorophenyl)-4-thiazolecarbohydrazide ≥95%
2-Amino-5-(4-chlorophenyl)-4-thiazolecarbohydrazide

Hazard: Irritant

Formula: C₁₀H₉ClN₄OS
Melting Pt: 257°C
MDL Number: MFCD06200937
CAS Number: 886361-54-2

Order Now

Specification Test Results

Melting point 257°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR