Ethyl-3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

043754-500MG 043754-1G 043754-5G
101862-400EA 375.38 USD
101862-400 101862-402 101862-404
Ethyl-3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₈BrClN₂O₂
MW: 303.54
Melting Pt: 144-146°C
MDL Number: MFCD06496148
CAS Number: 861208-16-4

Order Now

Specification Test Results

Melting point 144-146°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR