Methyl-4-bromo-1H-pyrrole-2-carboxylate ≥95%

Supplier: Matrix Scientific

043753-1G 043753-5G
101862-396EA 49.23 USD
101862-396 101862-398
Methyl-4-bromo-1H-pyrrole-2-carboxylate ≥95%
Methyl-4-bromo-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₆H₆BrNO₂
Melting Pt: 100…101 °C
MDL Number: MFCD00832855
CAS Number: 934-05-4

Order Now

Specification Test Results

Melting point 100-101°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR