2-(4-Methoxyphenyl)nicotinaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(4-Methoxyphenyl)pyridine-3-carbaldehyde
043748-500MG
043748-5G
101862-378EA
747.69
USD
101862-378
101862-382
2-(4-Methoxyphenyl)nicotinaldehyde ≥95%
2-(4-Methoxyphenyl)nicotinaldehyde
Hazard: Irritant
Formula:
C₁₃H₁₁NO₂ MW: 213.24 g/mol Melting Pt: 97…100 °C |
MDL Number:
MFCD06659546 CAS Number: 885949-59-7 |
Specification Test Results
Melting point | 97-100°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...