Methyl-4-(4-cyano-2-pyridinyl)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

043747-500MG 043747-1G 043747-5G
101862-372EA 556.92 USD
101862-372 101862-374 101862-376
Methyl-4-(4-cyano-2-pyridinyl)benzenecarboxylate ≥95%
Methyl-4-(4-cyano-2-pyridinyl)benzenecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₀N₂O₂
MW: 238.25 g/mol
Melting Pt: 170…172 °C
MDL Number: MFCD06200935
CAS Number: 886361-52-0

Order Now

Specification Test Results

Melting point 170-172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR