Methyl-4-[5-(chloromethyl)-2-pyridinyl]benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-[5-(chloromethyl)pyridin-2-yl]benzoate

043746-500MG 043746-1G 043746-5G
101862-366EA 556.92 USD
101862-366 101862-368 101862-370
Methyl-4-[5-(chloromethyl)-2-pyridinyl]benzenecarboxylate ≥95%
Methyl-4-[5-(chloromethyl)-2-pyridinyl]benzenecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₂ClNO₂
MW: 261.71 g/mol
Melting Pt: 80…83 °C
MDL Number: MFCD06200934
CAS Number: 886361-50-8

Order Now

Specification Test Results

Melting point 80-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR