Ethyl-1-(5-nitro-2-pyridinyl)-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

043744-1G 043744-5G
101862-358EA 232.31 USD
101862-358 101862-360
Ethyl-1-(5-nitro-2-pyridinyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(5-nitro-2-pyridinyl)-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₇N₃O₄
MW: 279.3 g/mol
Melting Pt: 67…70 °C
MDL Number: MFCD01308911
CAS Number: 300804-00-6

Order Now

Specification Test Results

Melting point 67-70°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR