1-[5-(4-Fluorophenyl)-2-thienyl]-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Acetyl-5-(4-fluorophenyl)thiophene

043737-500MG 043737-1G 043737-5G
101862-322EA 556.92 USD
101862-322 101862-324 101862-326
1-[5-(4-Fluorophenyl)-2-thienyl]-1-ethanone ≥95%
1-[5-(4-Fluorophenyl)-2-thienyl]-1-ethanone

Hazard: Irritant

Formula: C₁₂H₉FOS
MW: 220.27 g/mol
Melting Pt: 119…120 °C
MDL Number: MFCD06200932
CAS Number: 886361-46-2

Order Now

Specification Test Results

Melting point 119-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR