1-[5-(4-Chlorophenyl)-2-thienyl]-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Acetyl-5-(4-chlorophenyl)thiophene

043736-500MG 043736-1G 043736-5G
101862-316EA 375.38 USD
101862-316 101862-318 101862-320
1-[5-(4-Chlorophenyl)-2-thienyl]-1-ethanone ≥95%
1-[5-(4-Chlorophenyl)-2-thienyl]-1-ethanone

Hazard: Irritant

Formula: C₁₂H₉ClOS
MW: 236.72 g/mol
Melting Pt: 114…116 °C
MDL Number: MFCD01184547
CAS Number: 51335-90-1

Order Now

Specification Test Results

Melting point 114-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR