4'-Nitrobiphenyl-4-carboxaldehyde ≥95%
Supplier: Matrix Scientific
043735-500MG
043735-1G
043735-5G
101862-310EA
146.15
USD
101862-310
101862-312
101862-314
4'-Nitrobiphenyl-4-carboxaldehyde ≥95%
4'-Nitrobiphenyl-4-carboxaldehyde
Hazard: Irritant
Formula:
C₁₃H₉NO₃ MW: 227.22 g/mol Melting Pt: 117…120 °C |
MDL Number:
MFCD06200931 CAS Number: 98648-23-8 |
Specification Test Results
Melting point | 117-120°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...