1-[5-(4-Methoxyphenyl)-2-thienyl]-1-ethanone ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Acetyl-5-(4-methoxyphenyl)thiophene
043734-500MG
043734-1G
043734-5G
101862-304EA
375.38
USD
101862-304
101862-306
101862-308
1-[5-(4-Methoxyphenyl)-2-thienyl]-1-ethanone ≥95%
1-[5-(4-Methoxyphenyl)-2-thienyl]-1-ethanone
Hazard: Irritant
Formula:
C₁₃H₁₂O₂S MW: 232.3 g/mol Melting Pt: 156…157 °C |
MDL Number:
MFCD01860771 CAS Number: 13918-67-7 |
Specification Test Results
Melting point | 148-150°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...