2-Phenyl-1,3-thiazole-4-carbohydrazide ≥95%

Supplier: Matrix Scientific

043733-500MG 043733-1G 043733-5G
101862-298EA 278.46 USD
101862-298 101862-300 101862-302
2-Phenyl-1,3-thiazole-4-carbohydrazide ≥95%
2-Phenyl-1,3-thiazole-4-carbohydrazide

Hazard: Irritant

Formula: C₁₀H₉N₃OS
Melting Pt: 136…138 °C
MDL Number: MFCD00174011
CAS Number: 7113-12-4

Order Now

Specification Test Results

Melting point 136-138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR