5-(4-Chlorophenyl)-2-thiophenecarbaldehyde ≥95%
Supplier: Matrix Scientific
043732-500MG
043732-1G
043732-5G
101862-292EA
375.38
USD
101862-292
101862-294
101862-296
5-(4-Chlorophenyl)-2-thiophenecarbaldehyde ≥95%
5-(4-Chlorophenyl)-2-thiophenecarbaldehyde
Hazard: Irritant
Formula:
C₁₁H₇ClOS MW: 222.69 g/mol Melting Pt: 85…86 °C |
MDL Number:
MFCD01860767 CAS Number: 38401-71-7 |
Specification Test Results
Melting point | 85-86°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...