5-Phenyl-2-thiophenecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Phenylthiophene-2-carbaldehyde, 5-Phenylthiophene-2-carboxaldehyde

043731-500MG 043731-1G 043731-5G
101862-286EA 278.46 USD
101862-286 101862-288 101862-290
5-Phenyl-2-thiophenecarbaldehyde ≥95%
5-Phenyl-2-thiophenecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₈OS
Melting Pt: 92…94 °C
MDL Number: MFCD01151752
CAS Number: 19163-21-4

Order Now

Specification Test Results

Melting point 92-94°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR