Methyl-3-phenyl-1H-pyrazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Methoxycarbonyl)-3-phenyl-1H-pyrazole

043729-1G 043729-5G
101862-276EA 83.08 USD
101862-276 101862-278
Methyl-3-phenyl-1H-pyrazole-5-carboxylate ≥95%
Methyl-3-phenyl-1H-pyrazole-5-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂
MW: 202.21 g/mol
Melting Pt: 180…182 °C
MDL Number: MFCD00208415
CAS Number: 856061-37-5

Order Now

Specification Test Results

Melting point 180-182°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR