N-Methyl-N-(4-nitrophenyl)acetamide ≥95%
Supplier: Matrix Scientific
043728-500MG
043728-1G
043728-5G
101862-270EA
181.54
USD
101862-270
101862-272
101862-274
N-Methyl-N-(4-nitrophenyl)acetamide ≥95%
N-Methyl-N-(4-nitrophenyl)acetamide
Hazard: Irritant
Formula:
C₉H₁₀N₂O₃ Melting Pt: 153…154 °C |
MDL Number:
MFCD00035947 CAS Number: 121-95-9 |
Specification Test Results
Melting point | 153-154°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...