Pentafluorophenyl 2,4-dichlorobenzenesulfonate ≥95%

Supplier: Matrix Scientific

043726-500MG 043726-1G
101862-258EA 375.38 USD
101862-258 101862-260
Pentafluorophenyl 2,4-dichlorobenzenesulfonate ≥95%
Pentafluorophenyl 2,4-dichlorobenzenesulphonate

Hazard: Irritant

Formula: C₁₂H₃Cl₂F₅O₃S
Melting Pt: 79…81 °C
MDL Number: MFCD06659543
CAS Number: 885949-57-5

Order Now

Specification Test Results

Melting point 79-81°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR