Pentafluorophenyl 5-(dimethylamino)-1-naphthalenesulfonate ≥95%
Supplier: Matrix Scientific
043723-500MG
043723-5G
101862-240EA
375.38
USD
101862-240
101862-244
Pentafluorophenyl 5-(dimethylamino)-1-naphthalenesulfonate ≥95%
Pentafluorophenyl 5-(dimethylamino)-1-naphthalenesulphonate
Hazard: Irritant
Formula:
C₁₈H₁₂F₅NO₃S Melting Pt: 97-99°C |
MDL Number:
MFCD06659540 CAS Number: 663175-92-6 |
Specification Test Results
Melting point | 97-99°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...