Pentafluorophenyl 3,4-dichlorobenzenesulfonate ≥95%

Supplier: Matrix Scientific

043721-500MG 043721-1G 043721-5G
101862-228EA 375.38 USD
101862-228 101862-230 101862-232
Pentafluorophenyl 3,4-dichlorobenzenesulfonate ≥95%
Pentafluorophenyl 3,4-dichlorobenzenesulphonate

Hazard: Irritant

Formula: C₁₂H₃Cl₂F₅O₃S
Melting Pt: 76-78°C
MDL Number: MFCD06659538
CAS Number: 885949-53-1

Order Now

Specification Test Results

Melting point 76-78°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR