3-(3-Chlorophenyl)-3-(2,5-dimethyl-1H-pyrrol-1-yl)propanoic acid ≥95%

Supplier: Matrix Scientific

043718-500MG 043718-1G 043718-5G
101862-212EA 375.38 USD
101862-212 101862-214 101862-216
3-(3-Chlorophenyl)-3-(2,5-dimethyl-1H-pyrrol-1-yl)propanoic acid ≥95%
3-(3-Chlorophenyl)-3-(2,5-dimethyl-1H-pyrrol-1-yl)propanoic acid

Hazard: Irritant

Formula: C₁₅H₁₆ClNO₂
Melting Pt: 103…105 °C
MDL Number: MFCD04124841
CAS Number: 861207-88-7

Order Now

Specification Test Results

Melting point 103-105°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR