1-(4-Methoxyphenyl)-2-[(4-methylphenyl)sulfonyl]-1-ethanone ≥95%

Supplier: Matrix Scientific

043717-500MG 043717-1G 043717-5G
101862-206EA 375.38 USD
101862-206 101862-208 101862-210
1-(4-Methoxyphenyl)-2-[(4-methylphenyl)sulfonyl]-1-ethanone ≥95%
1-(4-Methoxyphenyl)-2-[(4-methylphenyl)sulfonyl]-1-ethanone

Hazard: Irritant

Formula: C₁₆H₁₆O₄S
Melting Pt: 130…131 °C
MDL Number: MFCD00220318
CAS Number: 86516-51-0

Order Now

Specification Test Results

Melting point 130-131°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR