4-Amino-6-chloropyrimidine-5-carboxaldehyde ≥95%

Supplier: Matrix Scientific

043716-500MG 043716-1G 043716-5G
101862-200EA 375.38 USD
101862-200 101862-202 101862-204
4-Amino-6-chloropyrimidine-5-carboxaldehyde ≥95%
4-Amino-6-chloropyrimidine-5-carboxaldehyde

Hazard: Irritant

Formula: C₅H₄ClN₃O
Melting Pt: >300 °C
MDL Number: MFCD03789626
CAS Number: 14160-93-1

Order Now

Specification Test Results

Melting point >300°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR