4-Chloro-6-morpholino-5-pyrimidinecarbaldehyde ≥95%

Supplier: Matrix Scientific

043715-500MG 043715-1G 043715-5G
101862-194EA 375.38 USD
101862-194 101862-196 101862-198
4-Chloro-6-morpholino-5-pyrimidinecarbaldehyde ≥95%
4-Chloro-6-morpholino-5-pyrimidinecarbaldehyde

Hazard: Irritant

Formula: C₉H₁₀ClN₃O₂
MW: 227.65 g/mol
Melting Pt: 94…96 °C
MDL Number: MFCD04124789
CAS Number: 54503-94-5

Order Now

Specification Test Results

Melting point 94-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR