4,6-Dichloropyrimidine-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

043714-1G 043714-5G 043714-25G
101862-188EA 23.08 USD
101862-188 101862-190 101862-192
4,6-Dichloropyrimidine-5-carbaldehyde ≥95%
4,6-Dichloropyrimidine-5-carbaldehyde

Hazard: Irritant-Harmful/Refrigerate/Avoid Skin Contact

Formula: C₅H₂Cl₂N₂O
MW: 175.99 g/mol
Melting Pt: 66…71 °C
MDL Number: MFCD02257701
CAS Number: 5305-40-8

Order Now

Specification Test Results

Melting point 70-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR