Ethyl-2-aminothiazole-4-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-aminothiazole-4-carboxylate
043713-5G
043713-25G
101862-184EA
20
USD
101862-184
101862-186
Ethyl-2-aminothiazole-4-carboxylate ≥95%
Ethyl-2-aminothiazole-4-carboxylate
Hazard: Irritant
Formula:
C₆H₈N₂O₂S Melting Pt: 175…177 °C |
MDL Number:
MFCD00619079 CAS Number: 5398-36-7 |
Specification Test Results
Melting point | 175-177°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...