2-(4-Benzylpiperazino)-2-methylpropanal ≥95%

Supplier: Matrix Scientific

043709-500MG 043709-1G 043709-5G
101862-170EA 556.92 USD
101862-170 101862-172 101862-174
2-(4-Benzylpiperazino)-2-methylpropanal ≥95%
2-(4-Benzylpiperazino)-2-methylpropanal

Hazard: Irritant

Formula: C₁₅H₂₂N₂O
MW: 246.36 g/mol
Melting Pt: 48…50 °C
MDL Number: MFCD05663785
CAS Number: 886360-87-8

Order Now

Specification Test Results

Melting point 48-50°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR