2-Hydroxy-4'-(methylsulfonyl)acetophenone ≥95%
Supplier: Matrix Scientific
043708-500MG
043708-1G
043708-5G
101862-164EA
130.77
USD
101862-164
101862-166
101862-168
2-Hydroxy-4'-(methylsulfonyl)acetophenone ≥95%
2-Hydroxy-4'-(methylsulphonyl)acetophenone
Hazard: Irritant
Formula:
C₉H₁₀O₄S Melting Pt: 136…138 °C |
CAS Number:
197240-27-0 |
Specification Test Results
Melting point | 136-138°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...