4-(4-Benzylpiperazino)-3-nitrobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

043706-500MG 043706-1G 043706-5G
101862-152EA 278.46 USD
101862-152 101862-154 101862-156
4-(4-Benzylpiperazino)-3-nitrobenzenecarbaldehyde ≥95%
4-(4-Benzylpiperazino)-3-nitrobenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₈H₁₉N₃O₃
MW: 325.37 g/mol
Melting Pt: 99…101 °C
MDL Number: MFCD00954856
CAS Number: 301193-53-3

Order Now

Specification Test Results

Melting point 99-101°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR