2-Methyl-1,3-benzoxazol-6-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methylbenzo[d]oxazol-6-amine

043697-500MG 043697-1G 043697-5G
101862-110EA 72.31 USD
101862-110 101862-112 101862-114
2-Methyl-1,3-benzoxazol-6-amine ≥95%
2-Methyl-1,3-benzoxazol-6-amine

Hazard: Irritant

Formula: C₁₁H₁₄N₂O
MW: 148.1619 g/mol
Melting Pt: 144…146 °C
MDL Number: MFCD03791181
CAS Number: 5676-60-8

Order Now

Specification Test Results

Melting point 144-146°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR