Methyl-2-amino-5-ethyl-1,3-thiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

043692-500MG 043692-1G 043692-5G
101862-086EA 556.92 USD
101862-086 101862-088 101862-090
Methyl-2-amino-5-ethyl-1,3-thiazole-4-carboxylate ≥95%
Methyl-2-amino-5-ethyl-1,3-thiazole-4-carboxylate

Hazard: Irritant

Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
Melting Pt: 120…122 °C
MDL Number: MFCD00728625
CAS Number: 28942-54-3

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR