Methyl-2-amino-5-isopropyl-1,3-thiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-5-isopropylthiazole-4-carboxylate

043691-500MG 043691-1G 043691-5G
101862-080EA 90.77 USD
101862-080 101862-082 101862-084
Methyl-2-amino-5-isopropyl-1,3-thiazole-4-carboxylate ≥95%
Methyl-2-amino-5-isopropyl-1,3-thiazole-4-carboxylate

Hazard: Irritant

Formula: C₈H₁₂N₂O₂S
MW: 200.26 g/mol
Melting Pt: 150…152 °C
MDL Number: MFCD01924792
CAS Number: 81569-25-7

Order Now

Specification Test Results

Melting point 150-152°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR