2-Nitro-5-piperidinobenzenecarboxamide ≥95%

Supplier: Matrix Scientific

043683-500MG 043683-1G
101862-048EA 556.92 USD
101862-048 101862-050
2-Nitro-5-piperidinobenzenecarboxamide ≥95%
2-Nitro-5-piperidinobenzenecarboxamide

Hazard: Irritant

Formula: C₁₂H₁₅N₃O₃
Melting Pt: 205…207 °C
MDL Number: MFCD03791176
CAS Number: 421558-77-2

Order Now

Specification Test Results

Melting point 205-207°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR