1-(6,7-Dichloro-2,3-dihydro-1,4-benzodioxin-5-yl)-1-ethanone ≥95%

Supplier: Matrix Scientific

043682-500MG 043682-1G 043682-5G
101862-042EA 278.46 USD
101862-042 101862-044 101862-046
1-(6,7-Dichloro-2,3-dihydro-1,4-benzodioxin-5-yl)-1-ethanone ≥95%
1-(6,7-Dichloro-2,3-dihydro-1,4-benzodioxin-5-yl)-1-ethanone

Hazard: Irritant

Formula: C₁₀H₈Cl₂O₃
MW: 247.08 g/mol
Melting Pt: 86…89 °C
MDL Number: MFCD03791175
CAS Number: 166816-11-1

Order Now

Specification Test Results

Melting point 86-89°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR