5-Nitro-1-benzothiophene-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Nitrobenzo[b]thiophene-2-carboxylic acid

043678-500MG 043678-1G 043678-5G
101862-024EA 375.38 USD
101862-024 101862-026 101862-028
5-Nitro-1-benzothiophene-2-carboxylic acid ≥95%
5-Nitro-1-benzothiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₅NO₄S
MW: 223.21 g/mol
Melting Pt: 239…241 °C
MDL Number: MFCD01159700
CAS Number: 6345-55-7

Order Now

Specification Test Results

Melting point 239-241°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR