3-Chloro-2-(1-methylhydrazino)-5-(trifluoromethyl)pyridine hydrochloride ≥95%

Supplier: Matrix Scientific

043673-500MG 043673-1G 043673-5G
101862-000EA 375.38 USD
101862-000 101862-002 101862-004
3-Chloro-2-(1-methylhydrazino)-5-(trifluoromethyl)pyridine hydrochloride ≥95%
3-Chloro-2-(1-methylhydrazino)-5-(trifluoromethyl)pyridine hydrochloride

Hazard: Irritant

Formula: C₇H₈Cl₂F₃N₃
MW: 262.06 g/mol
Melting Pt: 219…222 °C
MDL Number: MFCD03791252
CAS Number: 1049743-93-2

Order Now

Specification Test Results

Melting point 219-222°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR