N-[3-(Trifluoromethyl)phenyl]-1-hydrazinecarboxamide ≥95%

Supplier: Matrix Scientific

043672-500MG 043672-1G 043672-5G
101861-994EA 278.46 USD
101861-994 101861-996 101861-998
N-[3-(Trifluoromethyl)phenyl]-1-hydrazinecarboxamide ≥95%
N-[3-(Trifluoromethyl)phenyl]-1-hydrazinecarboxamide

Hazard: Irritant

Formula: C₈H₈F₃N₃O
Melting Pt: 92…98 °C
MDL Number: MFCD01869439
CAS Number: 448233-17-8

Order Now

Specification Test Results

Melting point 92-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR