Methyl-3-formyl-1H-indole-2-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 3-formyl-1H-indole-2-carboxylate
043669-500MG
043669-1G
043669-5G
101861-982EA
169.23
USD
101861-982
101861-984
101861-986
Methyl-3-formyl-1H-indole-2-carboxylate ≥95%
Methyl-3-formyl-1H-indole-2-carboxylate
Hazard: Irritant
Formula:
C₁₁H₉NO₃ MW: 203.2 g/mol Melting Pt: 206…208 °C |
MDL Number:
MFCD03848053 CAS Number: 18450-26-5 |
Specification Test Results
Melting point | 206-208°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...