4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]-3-methoxybenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

043667-500MG 043667-1G 043667-5G
101861-970EA 181.54 USD
101861-970 101861-972 101861-974
4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]-3-methoxybenzenecarbaldehyde ≥95%
4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]-3-methoxybenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₂H₁₀ClNO₃S
Melting Pt: 122…125 °C
MDL Number: MFCD01443861
CAS Number: 339018-41-6

Order Now

Specification Test Results

Melting point 122-125°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR