4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

043666-500MG 043666-1G 043666-5G
101861-964EA 278.46 USD
101861-964 101861-966 101861-968
4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]benzenecarbonitrile ≥95%
4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]benzenecarbonitrile

Hazard: Irritant

Formula: C₁₁H₇ClN₂OS
Melting Pt: 134…136 °C
MDL Number: MFCD01443858
CAS Number: 339018-30-3

Order Now

Specification Test Results

Melting point 134-136°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR