N-(4-Iodophenyl)cyclopropanecarboxamide ≥95%

Supplier: Matrix Scientific

043664-500MG 043664-1G 043664-5G
101861-952EA 181.54 USD
101861-952 101861-954 101861-956
N-(4-Iodophenyl)cyclopropanecarboxamide ≥95%
N-(4-Iodophenyl)cyclopropanecarboxamide

Hazard: Irritant

Formula: C₁₀H₁₀INO
Melting Pt: 214…216 °C
MDL Number: MFCD01327898
CAS Number: 23779-17-1

Order Now

Specification Test Results

Melting point 214-216°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR