Methyl-5-cyano-1,6-dihydro-1-(4-fluorophenyl)-4-methyl-6-pyridazinone-3-carboxylate ≥95%

Supplier: Matrix Scientific

043663-500MG 043663-1G 043663-5G
101861-946EA 1109.23 USD
101861-946 101861-948 101861-950
Methyl-5-cyano-1,6-dihydro-1-(4-fluorophenyl)-4-methyl-6-pyridazinone-3-carboxylate ≥95%
Methyl-5-cyano-1,6-dihydro-1-(4-fluorophenyl)-4-methyl-6-pyridazinone-3-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₀FN₃O₃
MW: 287.25 g/mol
Melting Pt: 163…165 °C
MDL Number: MFCD01315976
CAS Number: 339018-13-2

Order Now

Specification Test Results

Melting point 163-165°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR