2-(Aminooxy)-N-(5-iodo-2-pyridinyl)acetamide hydrochloride ≥95%
Supplier: Matrix Scientific
043662-500MG
043662-1G
043662-5G
101861-940EA
1109.23
USD
101861-940
101861-942
101861-944
2-(Aminooxy)-N-(5-iodo-2-pyridinyl)acetamide hydrochloride ≥95%
2-(Aminooxy)-N-(5-iodo-2-pyridinyl)acetamide hydrochloride
Hazard: Irritant
Formula:
C₇H₉ClIN₃O₂ Melting Pt: 197…199 °C |
MDL Number:
MFCD08056662 CAS Number: 321571-57-7 |
Specification Test Results
Melting point | 197-199°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...