N-(4-Chlorobenzyl)urea ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(4-Chlorobenzyl)urea
043661-500MG
043661-1G
043661-5G
101861-934EA
181.54
USD
101861-934
101861-936
101861-938
N-(4-Chlorobenzyl)urea ≥95%
N-(4-Chlorobenzyl)urea
Hazard: Irritant
Formula:
C₈H₉ClN₂O MW: 184.63 g/mol Melting Pt: 198…200 °C |
MDL Number:
MFCD01314630 CAS Number: 65608-74-4 |
Specification Test Results
Melting point | 198-200°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...