N-(3,4-Dimethoxybenzyl)urea ≥95%
Supplier: Matrix Scientific
043660-500MG
043660-1G
043660-5G
101861-928EA
181.54
USD
101861-928
101861-930
101861-932
N-(3,4-Dimethoxybenzyl)urea ≥95%
N-(3,4-Dimethoxybenzyl)urea
Hazard: Irritant
Formula:
C₁₀H₁₄N₂O₃ Melting Pt: 183…185 °C |
MDL Number:
MFCD01314629 CAS Number: 65609-19-0 |
Specification Test Results
Melting point | 183-185°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...