Methyl-5-chloro-6-oxo-1,6-dihydro-3-pyridinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 5-chloro-6-hydroxynicotinate, Methyl 5-chloro-6-oxo-1,6-dihydropyridine-3-carboxylate
043658-500MG
043658-1G
043658-5G
101861-916EA
1249.23
USD
101861-916
101861-918
101861-920
Methyl-5-chloro-6-oxo-1,6-dihydro-3-pyridinecarboxylate ≥95%
Methyl-5-chloro-6-oxo-1,6-dihydro-3-pyridinecarboxylate
Hazard: Irritant
Formula:
C₇H₆ClNO₃ MW: 187.58 g/mol Melting Pt: 220…222 °C |
MDL Number:
MFCD03844853 CAS Number: 316166-47-9 |
Specification Test Results
Melting point | 220-222°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...