1-(2-Chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-[(2-Chlorophenyl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid

043655-500MG 043655-1G 043655-5G
101861-900EA 278.46 USD
101861-900 101861-902 101861-904
1-(2-Chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid ≥95%
1-(2-Chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₀ClNO₃
MW: 263.68 g/mol
Melting Pt: 246…248 °C
MDL Number: MFCD01935944
CAS Number: 4399-77-3

Order Now

Specification Test Results

Melting point 246-248°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR