2-(6-Chloro-3-pyridazinyl)-2-(4-methylphenyl)acetonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Chloro-alpha-(4-methylphenyl)-3-pyridazineacetonitrile
043654-500MG
043654-1G
043654-5G
101861-894EA
181.54
USD
101861-894
101861-896
101861-898
2-(6-Chloro-3-pyridazinyl)-2-(4-methylphenyl)acetonitrile ≥95%
2-(6-Chloro-3-pyridazinyl)-2-(4-methylphenyl)acetonitrile
Hazard: Irritant
Formula:
C₁₃H₁₀ClN₃ MW: 243.70 g/mol Melting Pt: 133-135°C |
MDL Number:
MFCD00214777 CAS Number: 339008-32-1 |
Specification Test Results
Melting point | 133-135°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...