Methyl-3-{[3-(trifluoromethyl)benzyl]oxy}-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

043651-500MG 043651-1G 043651-5G
101861-876EA 181.54 USD
101861-876 101861-878 101861-880
Methyl-3-{[3-(trifluoromethyl)benzyl]oxy}-2-thiophenecarboxylate ≥95%
Methyl-3-{[3-(trifluoromethyl)benzyl]oxy}-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₁F₃O₃S
Melting Pt: 60…61 °C
MDL Number: MFCD00202083
CAS Number: 343375-98-4

Order Now

Specification Test Results

Melting point 60-61°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR