3-[(4-Methylbenzyl)oxy]-2-thiophenecarboxylic acid ≥95%

Supplier: Matrix Scientific

043650-500MG 043650-1G 043650-5G
101861-870EA 300 USD
101861-870 101861-872 101861-874
3-[(4-Methylbenzyl)oxy]-2-thiophenecarboxylic acid ≥95%
3-[(4-Methylbenzyl)oxy]-2-thiophenecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₂O₃S
Melting Pt: 139…141 °C
MDL Number: MFCD00215174
CAS Number: 343375-89-3

Order Now

Specification Test Results

Melting point 139-141°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR